siRNA Details
| Gene Symbol: GLTSCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015711.3 | NP_056526.3 | 14 | 3759 | ||
| XM_005258833.4 | XP_005258890.1 | 14 | 3802 | |||
| XM_006723180.3 | XP_006723243.1 | 10 | 2722 | |||
| XM_011526882.2 | XP_011525184.1 | 10 | 4769 | |||
| XM_011526883.2 | XP_011525185.1 | 13 | 3625 | |||
| GenBank mRNA | AF182077.1 | - | 9 | 3775 | ||

GLTSCR1
Genomic Map