siRNA Details
| Gene Symbol: CD53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000560.3 | NP_000551.1 | 5 | 544 | ||
| NM_001040033.1 | NP_001035122.1 | 6 | 549 | |||
| GenBank mRNA | AK313582.1 | - | 5 | 497 | ||
| BC035456.1 | - | 6 | 531 | |||
| BC040693.1 | - | 5 | 474 | 5498854 | ||
| BG536920.1 | - | 5 | 545 | |||
| BT019643.1 | - | 4 | 378 | |||
| CA453414.1 | - | 551 | ||||
| M37033.1 | - | 5 | 471 | |||
| M60871.1 | - | 5 | 451 | |||

CD53
Genomic Map