siRNA Details
| Gene Symbol: ADGRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271052.1 | NP_001257981.1 | 18 | 2380 | ||
| NM_013447.3 | NP_038475.2 | 20 | 2839 | |||
| XM_011527948.2 | XP_011526250.1 | 19 | 2468 | |||
| XM_011527949.2 | XP_011526251.1 | 19 | 2435 | |||
| XM_011527951.2 | XP_011526253.1 | 18 | 2321 | |||
| XM_011527952.2 | XP_011526254.1 | 18 | 2294 | |||
| XM_011527953.2 | XP_011526255.1 | 17 | 2275 | |||
| XM_011527954.2 | XP_011526256.1 | 16 | 3006 | |||
| XM_011527955.2 | XP_011526257.1 | 14 | 1885 | |||
| XM_017026726.1 | XP_016882215.1 | 18 | 2437 | |||
| XM_017026727.1 | XP_016882216.1 | 17 | 2114 | |||
| GenBank mRNA | AF114491.1 | - | 19 | 2452 | ||
| AK024426.1 | - | 10 | 1208 | |||
| AK024454.1 | - | 5 | 421 | |||
| AK222891.1 | - | 19 | 2437 | |||
| AK291776.1 | - | 19 | 2585 | |||
| AK298700.1 | - | 18 | 2296 | |||
| BC034949.1 | - | 15 | 2586 | |||
| BC053933.1 | - | 15 | 2579 | |||
| BC082991.1 | - | 19 | 2396 | |||
| BC127004.1 | - | 20 | 2835 | 40108332 | ||
| BC127005.1 | - | 20 | 2835 | 40108333 | ||
| CR936819.1 | - | 14 | 1816 | |||
More Information
Set Membership: |
|

ADGRE2
Genomic Map