siRNA Details
| Gene Symbol: UBQLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017481.2 | NP_059509.1 | 2 | 1066 | ||
| XM_011520144.2 | XP_011518446.1 | 2 | 1167 | |||
| GenBank mRNA | AF230481.1 | - | 2 | 1073 | ||
| BC036743.1 | - | 2 | 1081 | 5167911 | ||

UBQLN3
Genomic Map