siRNA Details
| Gene Symbol: IL21R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021798.3 | NP_068570.1 | 4 | 373 | ||
| NM_181078.2 | NP_851564.1 | 4 | 726 | |||
| NM_181079.4 | NP_851565.4 | 5 | 867 | |||
| XM_011545857.2 | XP_011544159.1 | 5 | 903 | |||
| XM_011545858.2 | XP_011544160.1 | 5 | 867 | |||
| XM_017023257.1 | XP_016878746.1 | 5 | 856 | |||
| GenBank mRNA | AF254067.1 | - | 4 | 321 | ||
| AF269133.1 | - | 4 | 389 | |||
| AK292663.1 | - | 4 | 726 | |||
| AK312825.1 | - | 4 | 715 | |||
| AY358826.1 | - | 4 | 356 | |||
| BC004348.1 | - | 4 | 338 | 3634520 | ||
| BQ215949.1 | - | 4 | 386 | |||
| BX401767.2 | - | 4 | 486 | |||

IL21R
Genomic Map