siRNA Details
| Gene Symbol: ANO7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001666.3 | NP_001001666.1 | 4 | 533 | ||
| NM_001001891.3 | NP_001001891.2 | 4 | 536 | |||
| XM_011511263.2 | XP_011509565.1 | 4 | 484 | |||
| XM_011511267.2 | XP_011509569.1 | 4 | 492 | |||
| XM_017004229.1 | XP_016859718.1 | 4 | 494 | |||
| XM_017004230.1 | XP_016859719.1 | 3 | 387 | |||
| GenBank mRNA | AY617079.1 | - | 4 | 537 | ||
| AY617080.1 | - | 4 | 534 | |||

ANO7
Genomic Map