siRNA Details
| Gene Symbol: PNPLA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256007.2 | NP_001242936.1 | 9 | 2128 | ||
| NM_001256008.2 | NP_001242937.1 | 8 | 2082 | |||
| NM_001256010.2 | NP_001242939.1 | 8 | 1955 | |||
| NM_001256011.2 | NP_001242940.1 | 9 | 1816 | |||
| NM_015723.4 | NP_056538.1 | 10 | 2240 | |||
| XM_011516274.2 | XP_011514576.1 | 9 | 2957 | |||
| GenBank mRNA | AB041261.1 | - | 8 | 1908 | ||
| AF263613.1 | - | 9 | 1934 | |||
| AK002115.1 | - | 5 | 428 | |||
| AK024335.1 | - | 9 | 1816 | |||
| AK291755.1 | - | 9 | 1945 | |||
| BC005203.1 | - | 5 | 394 | |||
| BC032999.2 | - | 10 | 2116 | 4829433 | ||
| BX647865.1 | - | 8 | 1801 | |||

PNPLA8
Genomic Map