siRNA Details
| Gene Symbol: CD70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252.4 | NP_001243.1 | 3 | 667 | ||
| GenBank mRNA | BC000725.2 | - | 3 | 613 | 3506629 | |
| BT007211.1 | - | 3 | 517 | |||
| L08096.1 | - | 3 | 667 | |||
| S69339.1 | - | 3 | 654 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD70| Gene Symbol: CD70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252.4 | NP_001243.1 | 3 | 667 | ||
| GenBank mRNA | BC000725.2 | - | 3 | 613 | 3506629 | |
| BT007211.1 | - | 3 | 517 | |||
| L08096.1 | - | 3 | 667 | |||
| S69339.1 | - | 3 | 654 | |||