siRNA Details
| Gene Symbol: CD70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252.4 | NP_001243.1 | 1 | 217 | ||
| XM_017027523.1 | XP_016883012.1 | 1 | 179 | |||
| GenBank mRNA | AK298467.1 | - | 1 | 179 | ||
| BC000725.2 | - | 1 | 163 | 3506629 | ||
| BT007211.1 | - | 1 | 67 | |||
| L08096.1 | - | 1 | 217 | |||
| S69339.1 | - | 1 | 204 | |||

CD70
Genomic Map