siRNA Details
| Gene Symbol: COPS7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164093.1 | NP_001157565.1 | 3 | 642 | ||
| NM_001164094.1 | NP_001157566.1 | 4 | 507 | |||
| NM_001164095.2 | NP_001157567.1 | 4 | 578 | |||
| NM_016319.3 | NP_057403.1 | 4 | 480 | |||
| XM_005253694.2 | XP_005253751.1 | 4 | 405 | |||
| GenBank mRNA | AB014764.1 | - | 3 | 268 | ||
| AB033603.1 | - | 4 | 345 | |||
| AF193844.1 | - | 3 | 268 | |||
| AF210052.1 | - | 4 | 409 | |||
| AK001318.1 | - | 4 | 392 | |||
| AK021721.1 | - | 4 | 445 | |||
| AK022488.1 | - | 4 | 387 | |||
| AK292621.1 | - | 3 | 638 | |||
| BC011789.2 | - | 4 | 368 | 4025790 | ||
| BC093015.1 | - | 4 | 419 | 30344094 | ||
| DA043679.1 | - | 4 | 507 | |||
| DA365275.1 | - | 4 | 452 | |||

COPS7A
Genomic Map