siRNA Details
| Gene Symbol: TAS2R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016943.2 | NP_058639.1 | 1 | 504 | ||
| GenBank mRNA | BC069337.1 | - | 1 | 442 | 7262266 | |
| BC095523.1 | - | 1 | 504 | 6971936 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS2R3| Gene Symbol: TAS2R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016943.2 | NP_058639.1 | 1 | 504 | ||
| GenBank mRNA | BC069337.1 | - | 1 | 442 | 7262266 | |
| BC095523.1 | - | 1 | 504 | 6971936 | ||