siRNA Details
| Gene Symbol: TAS2R7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023919.2 | NP_076408.1 | 1 | 138 | ||
| GenBank mRNA | BC069091.1 | - | 1 | 81 | 7216905 | |
| BC093992.1 | - | 1 | 138 | 7939837 | ||
| BC113561.1 | - | 1 | 138 | 8322613 | ||

TAS2R7
Genomic Map