siRNA Details
| Gene Symbol: TRAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317747.1 | NP_001304676.1 | 4 | 353 | ||
| NM_016388.3 | NP_057472.2 | 5 | 464 | |||
| GenBank mRNA | AF161547.1 | - | 5 | 379 | ||
| AJ224878.1 | - | 5 | 363 | |||
| AJ240085.1 | - | 4 | 279 | |||
| AK291991.1 | - | 5 | 464 | |||
| BC025713.1 | - | 5 | 370 | 5227396 | ||
| BX415345.2 | - | 5 | 372 | |||

TRAT1
Genomic Map