siRNA Details
| Gene Symbol: TRAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016388.3 | NP_057472.2 | 2, 3 | 350 | ||
| GenBank mRNA | AF161547.1 | - | 2, 3 | 265 | ||
| AJ224878.1 | - | 2, 3 | 249 | |||
| AK291991.1 | - | 2, 3 | 350 | |||
| BC025713.1 | - | 2, 3 | 256 | 5227396 | ||
| BX415345.2 | - | 2, 3 | 258 | |||

TRAT1
Genomic Map