siRNA Details
| Gene Symbol: SHANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016148.3 | NP_057232.2 | 23 | 6429 | ||
| XM_006723233.3 | XP_006723296.1 | 24 | 6827 | |||
| XM_011527013.2 | XP_011525315.1 | 25 | 6851 | |||
| XM_011527014.2 | XP_011525316.1 | 23 | 6800 | |||
| GenBank mRNA | AF163302.1 | - | 23 | 6429 | ||
| AF226728.1 | - | 9 | 4571 | |||

SHANK1
Genomic Map