siRNA Details
| Gene Symbol: CD79B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK222954.1 | - | 6 | 1158 | ||
| AK223210.1 | - | 6 | 1163 | |||
| AX775815.1 | - | 6 | 1154 | |||
| AX775817.1 | - | 6 | 1151 | |||
| BC002975.1 | - | 6 | 1138 | 3542354 | ||
| BC032651.1 | - | 6 | 1121 | 5504755 | ||
| M89957.1 | - | 6 | 1179 | |||

CD79B
Genomic Map