siRNA Details
| Gene Symbol: ISOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016048.2 | NP_057132.2 | 4 | 745 | ||
| GenBank mRNA | AF151869.1 | - | 4 | 744 | ||
| AK025787.1 | - | 4 | 743 | |||
| BC008367.1 | - | 4 | 723 | 3355904 | ||
| BC014105.2 | - | 4 | 733 | 4129769 | ||

ISOC1
Genomic Map