siRNA Details
| Gene Symbol: CD81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297649.1 | NP_001284578.1 | 6 | 620 | ||
| NM_004356.3 | NP_004347.1 | 6 | 738 | |||
| XM_011520492.2 | XP_011518794.1 | 6 | 972 | |||
| XM_017018598.1 | XP_016874087.1 | 6 | 481 | |||
| GenBank mRNA | AB209380.1 | - | 2 | 3753 | ||
| BC002978.1 | - | 6 | 545 | 3510634 | ||
| BC093047.1 | - | 6 | 736 | 30398483 | ||
| BG397506.1 | - | 7 | 608 | |||
| BT019507.1 | - | 6 | 505 | |||
| BT019508.1 | - | 6 | 505 | |||
| M33680.1 | - | 6 | 743 | |||

CD81
Genomic Map