siRNA Details
| Gene Symbol: BOLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321025.1 | NP_001307954.1 | 2 | 805 | ||
| NM_001321026.1 | NP_001307955.1 | 2 | 863 | |||
| NM_016074.4 | NP_057158.1 | 2 | 638 | |||
| XM_006711348.3 | XP_006711411.1 | 2 | 671 | |||
| GenBank mRNA | AF151901.1 | - | 2 | 574 | ||
| BC002771.2 | - | 2 | 598 | |||
| BC063405.1 | - | 2 | 598 | 5190565 | ||

BOLA1
Genomic Map