siRNA Details
| Gene Symbol: MRPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300848.1 | NP_001287777.1 | 4 | 661 | ||
| NM_015950.4 | NP_057034.2 | 4 | 661 | |||
| XM_005249161.4 | XP_005249218.1 | 4 | 482 | |||
| GenBank mRNA | AF132956.1 | - | 4 | 535 | ||
| AK301041.1 | - | 4 | 518 | |||
| AK314948.1 | - | 4 | 519 | |||
| BC013685.1 | - | 4 | 506 | 3862065 | ||
| BC020212.1 | - | 4 | 506 | 4641018 | ||
| BM563754.1 | - | 627 | ||||

MRPL2
Genomic Map