siRNA Details
| Gene Symbol: MRPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015956.2 | NP_057040.2 | 5 | 636 | ||
| NM_146387.1 | NP_666499.1 | 6 | 500 | |||
| NM_146388.1 | NP_666500.1 | 5 | 636 | |||
| XM_011528045.2 | XP_011526347.1 | 5 | 631 | |||
| GenBank mRNA | AB049635.1 | - | 5 | 351 | ||
| AF132962.1 | - | 5 | 597 | |||
| AY007152.1 | - | 4 | 289 | |||
| BC000756.2 | - | 5 | 590 | 2819730 | ||
| BC009858.2 | - | 5 | 377 | 3928448 | ||
| BG758801.1 | - | 6 | 563 | |||
| BM550378.1 | - | 6 | 498 | |||

MRPL4
Genomic Map