siRNA Details
| Gene Symbol: MRPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015956.2 | NP_057040.2 | 9 | 1123 | ||
| NM_146387.1 | NP_666499.1 | 10 | 987 | |||
| NM_146388.1 | NP_666500.1 | 8 | 2054 | |||
| GenBank mRNA | AB049635.1 | - | 9 | 838 | ||
| AF132962.1 | - | 9 | 1084 | |||
| AY007152.1 | - | 8 | 776 | |||
| BC009858.2 | - | 9 | 864 | 3928448 | ||

MRPL4
Genomic Map