siRNA Details
| Gene Symbol: NDUFA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015965.6 | NP_057049.5 | 5 | 516 | ||
| GenBank mRNA | AF261134.1 | - | 5 | 947 | ||
| AF286697.1 | - | 5 | 494 | |||
| BC000589.2 | - | 5 | 495 | |||
| BC009189.2 | - | 5 | 503 | 3957082 | ||
| BG705222.1 | - | 5 | 505 | |||
| BP230223.1 | - | 5 | 496 | |||

NDUFA13
Genomic Map