siRNA Details
| Gene Symbol: YBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015982.3 | NP_057066.2 | 9 | 1270 | ||
| XM_017024713.1 | XP_016880202.1 | 10 | 1360 | |||
| GenBank mRNA | AB208818.1 | - | 6 | 4251 | ||
| AF096834.1 | - | 9 | 1268 | |||
| AK092747.1 | - | 8 | 1887 | |||
| BC033800.1 | - | 9 | 1233 | 5171696 | ||
| BC047760.1 | - | 9 | 1234 | 5742121 | ||

YBX2
Genomic Map