siRNA Details
| Gene Symbol: UTP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016001.2 | NP_057085.2 | 7 | 1008 | ||
| XM_006721930.3 | XP_006721993.2 | 7 | 987 | |||
| XM_011524870.2 | XP_011523172.1 | 7 | 987 | |||
| GenBank mRNA | AF151806.1 | - | 7 | 994 | ||
| AK026355.1 | - | 8 | 757 | |||
| AK314399.1 | - | 7 | 987 | |||
| AY007138.1 | - | 5 | 478 | |||
| BC025276.1 | - | 7 | 984 | 4861495 | ||

UTP18
Genomic Map