siRNA Details
| Gene Symbol: NDUFAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016013.3 | NP_057097.2 | 5 | 1321 | ||
| NR_045620.1 | 6 | 1575 | ||||
| XM_006720555.2 | XP_006720618.1 | 5 | 1251 | |||
| XM_011521658.1 | XP_011519960.1 | 5 | 1185 | |||
| XM_011521659.2 | XP_011519961.1 | 5 | 1335 | |||
| GenBank mRNA | AF151823.1 | - | 5 | 1284 | ||
| AK310983.1 | - | 6 | 1575 | |||
| BC000780.2 | - | 5 | 1305 | 3504355 | ||
| BG192055.1 | - | 3 | 353 | |||

NDUFAF1
Genomic Map