siRNA Details
| Gene Symbol: COQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016035.4 | NP_057119.2 | 6 | 895 | ||
| XM_017014792.1 | XP_016870281.1 | 4 | 690 | |||
| GenBank mRNA | AF151850.1 | - | 6 | 866 | ||
| AK025376.1 | - | 6 | 809 | |||
| AK095083.1 | - | 2 | 2360 | |||
| AK128853.1 | - | 1 | 4559 | |||
| AK313650.1 | - | 6 | 878 | |||
| BC011895.2 | - | 6 | 565 | 3944239 | ||
| EU216419.1 | - | 6 | 877 | |||
| EU216420.1 | - | 6 | 795 | |||
| EU216421.1 | - | 6 | 629 | |||
| EU216422.1 | - | 6 | 567 | |||
| EU216423.1 | - | 5 | 491 | |||

COQ4
Genomic Map