siRNA Details
| Gene Symbol: CDC20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001255.2 | NP_001246.2 | 5, 6 | 653 | ||
| GenBank mRNA | AF099644.1 | - | 4, 5 | 596 | ||
| AK225504.1 | - | 5, 6 | 632 | |||
| AK302539.1 | - | 3, 4 | 885 | |||
| AK303844.1 | - | 6, 7 | 608 | |||
| AK312780.1 | - | 5, 6 | 632 | |||
| BC000624.2 | - | 4, 5 | 660 | 3346682 | ||
| BC001088.2 | - | 5, 6 | 653 | 2964686 | ||
| BC006272.2 | - | 4, 5 | 595 | 3948819 | ||
| BC009425.1 | - | 5, 6 | 635 | 2959460 | ||
| BC009426.1 | - | 5, 6 | 635 | 2959596 | ||
| BC010044.2 | - | 5, 6 | 644 | 3634656 | ||
| BC012803.2 | - | 5, 6 | 635 | 2959588 | ||
| BC012827.2 | - | 5, 6 | 653 | 2964686 | ||
| BC013303.2 | - | 5, 6 | 635 | 2959588 | ||
| BC015998.1 | - | 5, 6 | 631 | 4306426 | ||
| BC024257.1 | - | 5, 6 | 635 | 2959460 | ||
| BC031294.1 | - | 5, 6 | 635 | 2959487 | ||
| BC110321.1 | - | 4, 5 | 616 | 5496957 | ||
| BT007388.1 | - | 4, 5 | 552 | |||
| U05340.1 | - | 5, 6 | 662 | |||

CDC20
Genomic Map