siRNA Details
| Gene Symbol: IER3IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016097.4 | NP_057181.1 | 2 | 233 | ||
| GenBank mRNA | AF125100.1 | - | 2 | 216 | ||
| AF164798.1 | - | 2 | 202 | |||
| AF371963.1 | - | 2 | 206 | |||
| AK027108.1 | - | 2 | 223 | |||
| BC010888.1 | - | 2 | 199 | 4285974 | ||
| BC017391.1 | - | 2 | 166 | 4133818 | ||
| BM829309.1 | - | 2 | 232 | |||

IER3IP1
Genomic Map