siRNA Details
| Gene Symbol: SAR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033503.2 | NP_001028675.1 | 7, 8 | 744 | ||
| NM_016103.3 | NP_057187.1 | 6, 7 | 625 | |||
| XM_017009521.1 | XP_016865010.1 | 6, 7 | 632 | |||
| GenBank mRNA | AF087850.1 | - | 6, 7 | 662 | ||
| AF092130.1 | - | 6, 7 | 641 | |||
| AK056821.1 | - | 3, 4 | 2459 | |||
| AL512710.1 | - | 6, 7 | 1660 | |||
| BC002847.2 | - | 7, 8 | 675 | 3636540 | ||
| BC093034.1 | - | 6, 7 | 625 | 30528518 | ||

SAR1B
Genomic Map