siRNA Details
| Gene Symbol: ASB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201965.1 | NP_001188894.1 | 7 | 1494 | ||
| NM_016115.4 | NP_057199.1 | 8 | 1328 | |||
| NM_145863.2 | NP_665862.1 | 7 | 1119 | |||
| GenBank mRNA | AB209795.1 | - | 8 | 1227 | ||
| AF156778.1 | - | 7 | 1126 | |||
| AK000985.1 | - | 8 | 1254 | |||
| AK001283.1 | - | 7 | 1052 | |||
| AK056069.1 | - | 7 | 1496 | |||
| AK222595.1 | - | 7 | 1138 | |||
| AK222715.1 | - | 7 | 1110 | |||
| AK303133.1 | - | 6 | 1011 | |||
| BC006488.2 | - | 7 | 1140 | 3537359 | ||
| BC009569.1 | - | 8 | 1186 | 4052577 | ||
| Gene Symbol: GPR75-ASB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164165.1 | NP_001157637.1 | 8 | 1345 | ||
| GenBank mRNA | BC110915.1 | - | 8 | 1267 | 5764788 | |

ASB3
Genomic Map