siRNA Details
| Gene Symbol: CEP83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042399.1 | NP_001035858.1 | 5 | 1039 | ||
| NM_016122.2 | NP_057206.2 | 6 | 1092 | |||
| XM_006719437.2 | XP_006719500.1 | 5 | 1005 | |||
| XM_011538424.1 | XP_011536726.1 | 7 | 1138 | |||
| XM_011538427.2 | XP_011536729.1 | 6 | 864 | |||
| XM_011538429.2 | XP_011536731.1 | 2 | 335 | |||
| XM_017019385.1 | XP_016874874.1 | 6 | 1086 | |||
| XM_017019386.1 | XP_016874875.1 | 6 | 1086 | |||
| XM_017019387.1 | XP_016874876.1 | 5 | 843 | |||
| XM_017019388.1 | XP_016874877.1 | 5 | 2708 | |||
| XM_017019389.1 | XP_016874878.1 | 7 | 918 | |||
| GenBank mRNA | AF155115.1 | - | 6 | 808 | ||
| AK056316.1 | - | 5 | 935 | |||
| BC107746.1 | - | 3 | 353 | |||
| BC125086.1 | - | 5 | 729 | |||
| BC125087.1 | - | 6 | 642 | 40117807 | ||

CEP83
Genomic Map