siRNA Details
| Gene Symbol: DBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016216.3 | NP_057300.2 | 8 | 1195 | ||
| GenBank mRNA | AF180919.2 | - | 8 | 1149 | ||
| AK000116.1 | - | 3 | 672 | |||
| AK094147.1 | - | 9 | 1080 | |||
| BC009472.1 | - | 8 | 1123 | 3503627 | ||

DBR1
Genomic Map