siRNA Details
| Gene Symbol: AADAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286682.1 | NP_001273611.1 | 11, 12 | 1156 | ||
| NM_001286683.1 | NP_001273612.1 | 11, 12 | 1229 | |||
| NM_016228.3 | NP_057312.1 | 10, 11 | 1362 | |||
| NM_182662.1 | NP_872603.1 | 11, 12 | 1144 | |||
| XM_006714231.2 | XP_006714294.1 | 12, 13 | 1286 | |||
| XM_011532020.2 | XP_011530322.1 | 8, 9 | 919 | |||
| GenBank mRNA | AF481738.1 | - | 10, 11 | 1363 | ||
| AK055952.1 | - | 11, 12 | 1144 | |||
| AK314729.1 | - | 11, 12 | 1231 | |||
| BC031068.1 | - | 11, 12 | 1149 | 5286180 | ||

AADAT
Genomic Map