siRNA Details
| Gene Symbol: HSD17B14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016246.2 | NP_057330.2 | 7 | 784 | ||
| XM_005258969.3 | XP_005259026.1 | 7 | 793 | |||
| XM_005258970.3 | XP_005259027.1 | 6 | 703 | |||
| GenBank mRNA | AF126781.1 | - | 7 | 630 | ||
| AK307437.1 | - | 7 | 593 | |||
| AY358430.1 | - | 7 | 623 | |||
| BC006283.2 | - | 7 | 577 | 4026397 | ||
| BC006294.1 | - | 7 | 585 | 4025809 | ||

HSD17B14
Genomic Map