siRNA Details
| Gene Symbol: HSPA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016299.3 | NP_057383.2 | 8 | 884 | ||
| GenBank mRNA | AF112210.1 | - | 8 | 703 | ||
| AF143723.1 | - | 8 | 693 | |||
| AK225265.1 | - | 8 | 728 | |||
| AK225651.1 | - | 8 | 728 | |||
| AK226065.1 | - | 1 | 3557 | |||
| AK292323.1 | - | 8 | 692 | |||
| BC119690.1 | - | 8 | 703 | 40116697 | ||

HSPA14
Genomic Map