siRNA Details
Gene Symbol: TCEAL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006612.1 | NP_001006613.1 | 3 | 560 | ||
NM_001006613.1 | NP_001006614.1 | 2 | 484 | |||
NM_001006614.1 | NP_001006615.1 | 2 | 481 | |||
NM_016303.2 | NP_057387.1 | 3 | 563 | |||
GenBank mRNA | AF125535.1 | - | 3 | 521 | ||
AK312190.1 | - | 3 | 509 | |||
AL833152.1 | - | 1 | 1427 | |||
BC023544.2 | - | 3 | 514 | 4122244 |
