siRNA Details
| Gene Symbol: TCEAL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006612.1 | NP_001006613.1 | 3 | 507 | ||
| NM_001006613.1 | NP_001006614.1 | 2 | 431 | |||
| NM_001006614.1 | NP_001006615.1 | 2 | 428 | |||
| NM_016303.2 | NP_057387.1 | 3 | 510 | |||
| GenBank mRNA | AF125535.1 | - | 3 | 468 | ||
| AK312190.1 | - | 3 | 456 | |||
| AL833152.1 | - | 1 | 1374 | |||
| BC023544.2 | - | 3 | 461 | 4122244 | ||

TCEAL9
Genomic Map