siRNA Details
| Gene Symbol: DDX47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016355.3 | NP_057439.2 | 9, 10 | 1065 | ||
| NM_201224.1 | NP_957518.1 | 8, 9 | 918 | |||
| GenBank mRNA | AF078843.1 | - | 8, 9 | 875 | ||
| AF190165.1 | - | 8, 9 | 1578 | |||
| AJ276704.1 | - | 9, 10 | 1043 | |||
| AK054574.1 | - | 9, 10 | 1050 | |||
| AK222940.1 | - | 9, 10 | 1024 | |||
| AK225144.1 | - | 9, 10 | 1035 | |||
| AL136666.1 | - | 9, 10 | 1069 | |||
| AL833469.1 | - | 6, 7 | 1938 | |||
| BC009379.2 | - | 9, 10 | 1028 | |||
| BC068009.1 | - | 9, 10 | 1047 | 6142837 | ||

DDX47
Genomic Map