siRNA Details
| Gene Symbol: DDX47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016355.3 | NP_057439.2 | 5 | 549 | ||
| NM_201224.1 | NP_957518.1 | 5 | 549 | |||
| GenBank mRNA | AF078843.1 | - | 5 | 506 | ||
| AF190165.1 | - | 5 | 513 | |||
| AJ276704.1 | - | 5 | 527 | |||
| AK054574.1 | - | 5 | 534 | |||
| AK222940.1 | - | 5 | 508 | |||
| AK225144.1 | - | 5 | 519 | |||
| AK310501.1 | - | 5 | 579 | |||
| AL136666.1 | - | 5 | 553 | |||
| AL833469.1 | - | 4 | 349 | |||
| BC009379.2 | - | 5 | 512 | |||
| BC068009.1 | - | 5 | 531 | 6142837 | ||
| EF036507.1 | - | 5 | 496 | |||

DDX47
Genomic Map