siRNA Details
| Gene Symbol: ACP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB030039.2 | - | 7 | 1096 | ||
| AB031478.1 | - | 7 | 1203 | |||
| AK000657.1 | - | 7 | 1029 | |||
| AY358552.1 | - | 7 | 1000 | |||
| BC009965.1 | - | 7 | 1272 | 4299943 | ||
| BC034686.1 | - | 7 | 1276 | 3463770 | ||
| CR457101.1 | - | 7 | 807 | |||
| KJ534759.1 | - | 7 | 828 | |||
| KJ535036.1 | - | 6 | 700 | |||

ACP6
Genomic Map