siRNA Details
| Gene Symbol: GP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083899.2 | NP_001077368.2 | 6 | 725 | ||
| NM_001256017.2 | NP_001242946.2 | 5 | 671 | |||
| NM_016363.5 | NP_057447.5 | 6 | 725 | |||
| GenBank mRNA | AB035073.1 | - | 6 | 709 | ||
| AB043819.1 | - | 6 | 725 | |||
| AB043820.1 | - | 5 | 671 | |||
| AB043821.1 | - | 6 | 725 | |||
| BC104832.1 | - | 6 | 717 | 8143835 | ||
| BC111963.1 | - | 6 | 717 | 8327431 | ||
| BQ216894.1 | - | 1 | 152 | |||

GP6
Genomic Map