siRNA Details
| Gene Symbol: RAB9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016370.2 | NP_057454.1 | 3 | 713 | ||
| GenBank mRNA | AB036693.1 | - | 3 | 659 | ||
| AK313425.1 | - | 3 | 608 | |||
| BC093756.1 | - | 1 | 397 | 7939601 | ||
| BC093758.1 | - | 1 | 397 | 7939603 | ||

RAB9B
Genomic Map