siRNA Details
| Gene Symbol: EMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286420.1 | NP_001273349.1 | 2 | 248 | ||
| NM_016454.3 | NP_057538.1 | 2 | 248 | |||
| GenBank mRNA | AF151018.1 | - | 2 | 201 | ||
| AK075227.1 | - | 2 | 227 | |||
| AK290524.1 | - | 2 | 232 | |||
| AK296184.1 | - | 2 | 228 | |||
| BC002583.2 | - | 2 | 213 | 3161718 | ||
| BG707804.1 | - | 2 | 253 | |||
| KU178640.1 | - | 2 | 130 | |||
| KU178641.1 | - | 2 | 130 | |||

EMC4
Genomic Map