siRNA Details
| Gene Symbol: EMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286420.1 | NP_001273349.1 | 3 | 410 | ||
| NM_016454.3 | NP_057538.1 | 3 | 410 | |||
| GenBank mRNA | AF151018.1 | - | 3 | 363 | ||
| AK075227.1 | - | 3 | 389 | |||
| AK290524.1 | - | 3 | 394 | |||
| AY336092.1 | - | 3 | 350 | |||
| BC002583.2 | - | 3 | 375 | 3161718 | ||
| BG707804.1 | - | 3 | 423 | |||
| KU178640.1 | - | 3 | 292 | |||
| KU178641.1 | - | 3 | 292 | |||

EMC4
Genomic Map