siRNA Details
| Gene Symbol: HGH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016458.3 | NP_057542.2 | 3 | 779 | ||
| GenBank mRNA | AK098226.1 | - | 4 | 473 | ||
| BC003035.1 | - | 3 | 779 | 2822563 | ||
| BC009915.2 | - | 3 | 779 | 2822563 | ||
| BC015471.1 | - | 4 | 310 | |||

HGH1
Genomic Map