siRNA Details
| Gene Symbol: SHISA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272065.1 | NP_001258994.1 | 6 | 1747 | ||
| NM_001272066.1 | NP_001258995.1 | 6 | 1734 | |||
| NM_001272067.1 | NP_001258996.1 | 6 | 1929 | |||
| NM_001272068.1 | NP_001258997.1 | 7 | 1966 | |||
| NM_001272082.1 | NP_001259011.1 | 4 | 1623 | |||
| NM_001272083.1 | NP_001259012.1 | 3 | 1797 | |||
| NM_016479.4 | NP_057563.3 | 6 | 1768 | |||
| XM_005265205.1 | XP_005265262.1 | 7 | 2016 | |||
| XM_006713188.2 | XP_006713251.1 | 6 | 1669 | |||
| XM_011533796.1 | XP_011532098.1 | 7 | 1758 | |||
| GenBank mRNA | AF151051.1 | - | 5 | 1391 | ||
| AF520698.1 | - | 6 | 1747 | |||
| AK027728.1 | - | 990 | ||||
| AK056328.1 | - | 6 | 1910 | |||
| AK075441.1 | - | 6 | 1683 | |||
| AK124652.1 | - | 7 | 1966 | |||
| AK298842.1 | - | 4 | 1398 | |||
| AL117413.1 | - | 3 | 1568 | |||
| AL832568.1 | - | 4 | 1430 | |||
| BC001463.2 | - | 4 | 1438 | 3138644 | ||
| BC020279.2 | - | 4 | 1388 | |||

SHISA5
Genomic Map