siRNA Details
| Gene Symbol: TMEM216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173990.2 | NP_001167461.1 | 4 | 533 | ||
| NM_001173991.2 | NP_001167462.1 | 4 | 533 | |||
| NM_016499.5 | NP_057583.2 | 4 | 514 | |||
| XM_005274039.3 | XP_005274096.1 | 5 | 1354 | |||
| XM_017017882.1 | XP_016873371.1 | 4 | 1220 | |||
| GenBank mRNA | AK303687.1 | - | 4 | 533 | ||
| BG759319.1 | - | 4 | 515 | |||
| KU159424.1 | - | 4 | 320 | |||

TMEM216
Genomic Map