siRNA Details
| Gene Symbol: PBDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300888.1 | NP_001287817.1 | 5 | 556 | ||
| NM_016500.3 | NP_057584.2 | 6 | 661 | |||
| GenBank mRNA | BC001220.1 | - | 6 | 661 | 3528586 | |
| BC051894.1 | - | 6 | 523 | 6528202 | ||
| CD704296.1 | - | 5 | 450 | |||

PBDC1
Genomic Map