siRNA Details
| Gene Symbol: MRPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016504.2 | NP_057588.1 | 2 | 141 | ||
| GenBank mRNA | AB049647.1 | - | 2 | 116 | ||
| AF151084.1 | - | 2 | 125 | |||
| AK023576.1 | - | 2 | 122 | |||
| AK315758.1 | - | 2 | 142 | |||
| BC001066.2 | - | 2 | 112 | 2822502 | ||
| BC021986.1 | - | 2 | 126 | 4095663 | ||
| BG829493.1 | - | 2 | 160 | |||
| BM840463.1 | - | 2 | 142 | |||

MRPL27
Genomic Map